Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fpr-rs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169140.1 | NP_001162611.1 | 1 | 546 | 90 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon |