Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmur2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022275.3 | NP_071611.3 | 1 - 2 | 830 | 89 | |
GenBank mRNA | AB041229.1 | - | 1 - 2 | 830 | 89 | |
AF242875.1 | - | 1 - 2 | 704 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Rattus norvegicus Probe spans exons |