Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053656.2 | NP_446108.2 | 4 - 5 | 458 | 67 | |
XM_006249515.2 | XP_006249577.1 | 4 - 5 | 651 | 67 | ||
XM_006249516.2 | XP_006249578.1 | 4 - 5 | 650 | 67 | ||
XM_006249517.2 | XP_006249579.1 | 4 - 5 | 613 | 67 | ||
XM_008769305.1 | XP_008767527.1 | 4 - 5 | 615 | 67 | ||
XM_008769306.1 | XP_008767528.1 | 4 - 5 | 327 | 67 | ||
GenBank mRNA | AF020756.1 | - | 4 - 5 | 439 | 67 | |
AF020757.1 | - | 4 - 5 | 485 | 67 | ||
AF020758.1 | - | 3 - 4 | 579 | 67 | ||
AF020759.1 | - | 4 - 5 | 420 | 67 | ||
AF064549.1 | - | 3 - 4 | 495 | 67 | ||
U14414.1 | - | 4 - 5 | 458 | 67 | ||
Y10473.1 | - | 4 - 5 | 458 | 67 | ||
Y10474.1 | - | 4 - 5 | 440 | 67 | ||
Y10475.1 | - | 4 - 5 | 494 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Detect Genomic DNA Rattus norvegicus Probe spans exons |