Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053656.2 | NP_446108.2 | 6 - 7 | 636 | 60 | |
XM_006249515.2 | XP_006249577.1 | 6 - 7 | 829 | 60 | ||
XM_006249516.2 | XP_006249578.1 | 6 - 7 | 828 | 60 | ||
XM_006249517.2 | XP_006249579.1 | 6 - 7 | 791 | 60 | ||
XM_008769306.1 | XP_008767528.1 | 6 - 7 | 505 | 60 | ||
XM_008769307.1 | XP_008767529.1 | 4 - 5 | 577 | 60 | ||
GenBank mRNA | AF020759.1 | - | 6 - 7 | 598 | 60 | |
AF064549.1 | - | 5 - 6 | 673 | 60 | ||
U14414.1 | - | 6 - 7 | 636 | 60 | ||
Y10475.1 | - | 6 - 7 | 672 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons |