Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cst6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133566.1 | NP_598250.1 | 2 - 3 | 642 | 79 | |
GenBank mRNA | AY046415.1 | - | 2 - 3 | 642 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |