Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apoe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270681.1 | NP_001257610.1 | 3 - 4 | 352 | 98 | |
NM_001270682.1 | NP_001257611.1 | 3 - 4 | 331 | 98 | ||
NM_001270683.1 | NP_001257612.1 | 3 - 4 | 318 | 98 | ||
NM_001270684.1 | NP_001257613.1 | 3 - 4 | 294 | 98 | ||
NM_138828.3 | NP_620183.2 | 3 - 4 | 312 | 98 | ||
GenBank mRNA | BC060313.1 | - | 3 - 4 | 285 | 6919048 | 98 |
BC086581.1 | - | 3 - 4 | 255 | 7307758 | 98 | |
FQ209426.1 | - | 3 - 4 | 283 | 98 | ||
FQ209457.1 | - | 3 - 4 | 280 | 98 | ||
FQ209625.1 | - | 3 - 4 | 281 | 98 | ||
FQ209879.1 | - | 2 - 3 | 228 | 98 | ||
FQ209926.1 | - | 3 - 4 | 281 | 98 | ||
FQ210262.1 | - | 3 - 4 | 281 | 98 | ||
FQ210410.1 | - | 3 - 4 | 281 | 98 | ||
FQ211250.1 | - | 3 - 4 | 282 | 98 | ||
FQ212760.1 | - | 3 - 4 | 281 | 98 | ||
FQ212769.1 | - | 3 - 4 | 281 | 98 | ||
FQ213060.1 | - | 3 - 4 | 277 | 98 | ||
FQ213396.1 | - | 3 - 4 | 277 | 98 | ||
FQ213533.1 | - | 3 - 4 | 284 | 98 | ||
FQ213898.1 | - | 3 - 4 | 273 | 98 | ||
FQ214072.1 | - | 3 - 4 | 281 | 98 | ||
FQ214282.1 | - | 3 - 4 | 281 | 98 | ||
FQ218386.1 | - | 3 - 4 | 280 | 98 | ||
FQ218473.1 | - | 3 - 4 | 310 | 98 | ||
FQ218485.1 | - | 3 - 4 | 281 | 98 | ||
FQ218540.1 | - | 3 - 4 | 281 | 98 | ||
FQ218699.1 | - | 3 - 4 | 283 | 98 | ||
FQ218898.1 | - | 3 - 4 | 283 | 98 | ||
FQ218953.1 | - | 2 - 3 | 337 | 98 | ||
FQ219009.1 | - | 3 - 4 | 281 | 98 | ||
FQ219124.1 | - | 2 - 3 | 241 | 98 | ||
FQ219144.1 | - | 3 - 4 | 281 | 98 | ||
FQ219202.1 | - | 2 - 3 | 232 | 98 | ||
FQ219319.1 | - | 3 - 4 | 281 | 98 | ||
FQ219432.1 | - | 3 - 4 | 281 | 98 | ||
FQ219448.1 | - | 3 - 4 | 283 | 98 | ||
FQ219490.1 | - | 3 - 4 | 281 | 98 | ||
FQ219516.1 | - | 3 - 4 | 281 | 98 | ||
FQ219534.1 | - | 3 - 4 | 281 | 98 | ||
FQ219700.1 | - | 3 - 4 | 281 | 98 | ||
FQ219711.1 | - | 3 - 4 | 281 | 98 | ||
FQ220125.1 | - | 3 - 4 | 277 | 98 | ||
FQ220532.1 | - | 3 - 4 | 277 | 98 | ||
FQ220683.1 | - | 3 - 4 | 281 | 98 | ||
FQ220752.1 | - | 3 - 4 | 312 | 98 | ||
FQ220879.1 | - | 3 - 4 | 285 | 98 | ||
FQ221856.1 | - | 3 - 4 | 281 | 98 | ||
FQ229186.1 | - | 3 - 4 | 281 | 98 | ||
FQ229530.1 | - | 2 - 3 | 232 | 98 | ||
FQ229565.1 | - | 3 - 4 | 277 | 98 | ||
FQ229759.1 | - | 3 - 4 | 281 | 98 | ||
FQ229950.1 | - | 3 - 4 | 281 | 98 | ||
FQ230084.1 | - | 3 - 4 | 281 | 98 | ||
FQ231375.1 | - | 3 - 4 | 310 | 98 | ||
J00705.1 | - | 2 - 3 | 239 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |