Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spink1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152936.1 | NP_690919.1 | 2 - 3 | 148 | 98 | |
GenBank mRNA | FQ209906.1 | - | 2 - 3 | 129 | 98 | |
FQ210000.1 | - | 2 - 3 | 162 | 98 | ||
FQ210029.1 | - | 2 - 3 | 161 | 98 | ||
FQ210138.1 | - | 2 - 3 | 161 | 98 | ||
FQ210344.1 | - | 2 - 3 | 161 | 98 | ||
FQ210442.1 | - | 2 - 3 | 161 | 98 | ||
FQ210447.1 | - | 2 - 3 | 161 | 98 | ||
FQ210766.1 | - | 2 - 3 | 163 | 98 | ||
FQ210836.1 | - | 2 - 3 | 163 | 98 | ||
FQ210933.1 | - | 2 - 3 | 163 | 98 | ||
FQ211214.1 | - | 2 - 3 | 165 | 98 | ||
FQ211434.1 | - | 2 - 3 | 147 | 98 | ||
FQ218107.1 | - | 2 - 3 | 163 | 98 | ||
FQ218122.1 | - | 2 - 3 | 162 | 98 | ||
FQ218165.1 | - | 2 - 3 | 163 | 98 | ||
FQ218208.1 | - | 2 - 3 | 159 | 98 | ||
FQ218422.1 | - | 2 - 3 | 161 | 98 | ||
M27883.1 | - | 2 - 3 | 148 | 98 |
Gene Symbol: LOC100911558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751771.3 | XP_003751819.1 | 2 - 3 | 498 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Rattus norvegicus Probe spans exons |