Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177426.1 | NP_803175.1 | 363 | 97 | ||
GenBank mRNA | FJ179402.1 | - | 363 | 97 | ||
FQ214985.1 | - | 5 - 6 | 396 | 97 | ||
FQ218547.1 | - | 5 - 6 | 429 | 97 | ||
FQ218596.1 | - | 5 - 6 | 396 | 97 | ||
FQ219560.1 | - | 5 - 6 | 396 | 97 | ||
J02592.1 | - | 3 - 4 | 267 | 97 | ||
M13590.1 | - | 4 - 5 | 293 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Inventoried Probe spans exons |