Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lenep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053614.1 | NP_446066.1 | 1 | 250 | 110 | |
GenBank mRNA | U15149.1 | - | 1 | 250 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Amplicon spans exons and probe does not span exons |