Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom2r18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173318.1 | NP_775440.1 | 1 | 150 | 80 | |
GenBank mRNA | AF016181.1 | - | 1 | 150 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon |