Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173526.1 | NP_001166997.1 | 4 - 5 | 941 | 70 | |
XM_008759380.1 | XP_008757602.1 | 6 - 7 | 2251 | 70 | ||
XM_008759381.1 | XP_008757603.1 | 7 - 8 | 2101 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |