Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdrd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191583.1 | NP_001178512.1 | 3 - 4 | 6285 | 101 | |
XM_006244608.2 | XP_006244670.1 | 3 - 4 | 6424 | 101 | ||
XM_006244609.2 | XP_006244671.1 | 3 - 4 | 6421 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus Probe spans exons |