Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191641.1 | NP_001178570.1 | 2 - 3 | 518 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |