Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ghdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191651.1 | NP_001178580.1 | 2 - 3 | 344 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |