Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Padi6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191766.1 | NP_001178695.1 | 4 - 5 | 412 | 65 | |
XM_008764232.1 | XP_008762454.1 | 4 - 5 | 462 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |