Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191830.1 | NP_001178759.1 | 1 - 2 | 270 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Protein Phosphatase 3' Most Recommended Assay Rattus norvegicus Inventoried Probe spans exons |