Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb5d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191890.1 | NP_001178819.1 | 1 - 2 | 160 | 93 | |
XM_006246767.2 | XP_006246829.1 | 2 - 3 | 212 | 93 | ||
GenBank mRNA | FQ223142.1 | - | 1 - 2 | 551 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |