Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Noto | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192014.1 | NP_001178943.1 | 1 - 2 | 384 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |