Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100359907 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195414.1 | NP_001182343.1 | 3 - 4 | 192 | 65 | |
GenBank mRNA | HM015624.1 | - | 3 - 4 | 192 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |