Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sftpd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012878.2 | NP_037010.1 | 3 - 4 | 450 | 85 | |
XM_008771065.1 | XP_008769287.1 | 3 - 4 | 453 | 85 | ||
GenBank mRNA | BC070507.1 | - | 4 - 5 | 459 | 7098194 | 85 |
M81231.1 | - | 4 - 5 | 457 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |