Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etv3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100992.2 | NP_001094462.2 | 4 - 5 | 740 | 89 | |
XM_006232728.2 | XP_006232790.1 | 4 - 5 | 1003 | 89 | ||
XM_006232729.2 | XP_006232791.1 | 4 - 5 | 1020 | 89 | ||
XM_006232730.2 | XP_006232792.1 | 5 - 6 | 671 | 89 | ||
XM_006232731.2 | XP_006232793.1 | 5 - 6 | 644 | 89 | ||
GenBank mRNA | BC127525.1 | - | 4 - 5 | 740 | 7114610 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |