Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsga10ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004278.3 | NP_001004278.2 | 2 - 3 | 302 | 67 | |
NM_001270736.1 | NP_001257665.1 | 2 - 3 | 302 | 67 | ||
NM_001270737.1 | NP_001257666.1 | 2 - 3 | 302 | 67 | ||
XM_006230847.2 | XP_006230909.1 | 2 - 3 | 577 | 67 | ||
XM_008760155.1 | XP_008758377.1 | 2 - 3 | 587 | 67 | ||
XM_008760156.1 | XP_008758378.1 | 2 - 3 | 586 | 67 | ||
XM_008760157.1 | XP_008758379.1 | 2 - 3 | 588 | 67 | ||
GenBank mRNA | AY690667.1 | - | 2 - 3 | 319 | 67 | |
BC097254.1 | - | 2 - 3 | 283 | 7453704 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |