Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Faim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014843.2 | NP_001014843.1 | 6 - 7 | 1141 | 102 | |
GenBank mRNA | BC088197.1 | - | 6 - 7 | 1141 | 7375085 | 102 |
FQ232848.1 | - | 3 - 4 | 585 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |