Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crisp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039393.2 | NP_001034482.2 | 2 - 3 | 238 | 94 | |
XM_008766935.1 | XP_008765157.1 | 2 - 3 | 238 | 94 | ||
GenBank mRNA | DQ200907.1 | - | 2 - 3 | 238 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Protease Rattus norvegicus Probe spans exons |