Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srxn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047858.3 | NP_001041323.3 | 1 - 2 | 450 | 90 | |
XM_008762308.1 | XP_008760530.1 | 2 - 3 | 245 | 90 | ||
GenBank mRNA | AY325205.1 | - | 2 - 3 | 245 | 90 | |
BC167743.1 | - | 1 - 2 | 203 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons |