Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnmb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104560.2 | NP_001098030.1 | 2 - 3 | 493 | 88 | |
GenBank mRNA | AB297662.1 | - | 2 - 3 | 494 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Ion Channel Rattus norvegicus Probe spans exons |