Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsga10ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270736.1 | NP_001257665.1 | 5 - 6 | 1198 | 84 | |
XM_006230847.2 | XP_006230909.1 | 5 - 6 | 1473 | 84 | ||
XM_008760155.1 | XP_008758377.1 | 5 - 6 | 1483 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |