Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr1i3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270838.1 | NP_001257767.1 | 7 - 8 | 986 | 87 | |
NM_001270840.1 | NP_001257769.1 | 7 - 8 | 983 | 87 | ||
NM_022941.4 | NP_075230.1 | 7 - 8 | 986 | 87 | ||
XM_006250261.2 | XP_006250323.1 | 7 - 8 | 1092 | 87 | ||
GenBank mRNA | AB104736.1 | - | 7 - 8 | 1039 | 87 | |
AB105072.1 | - | 6 - 7 | 1130 | 87 | ||
AB204897.1 | - | 6 - 7 | 834 | 87 | ||
AB204898.1 | - | 6 - 7 | 837 | 87 | ||
AF133094.1 | - | 6 - 7 | 837 | 87 | ||
AF133095.1 | - | 6 - 7 | 837 | 87 | ||
BC089763.1 | - | 7 - 8 | 983 | 7324318 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Nuclear Receptor Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |