Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rorb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270958.1 | NP_001257887.1 | 9 - 10 | 1850 | 72 | |
XM_002728869.3 | XP_002728915.1 | 9 - 10 | 3502 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Nuclear Receptor 3' Most Rattus norvegicus 1700 Probe spans exons |