Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgol2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271191.1 | NP_001258120.1 | 6 - 7 | 3325 | 107 | |
XM_006244988.2 | XP_006245050.1 | 7 - 8 | 3602 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons |