Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spock1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271297.1 | NP_001258226.1 | 869 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |