Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fzd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022623.2 | NP_072145.1 | 1 - 2 | 469 | 87 | |
GenBank mRNA | AF183910.1 | - | 1 - 2 | 500 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Rattus norvegicus 1700 Probe spans exons |