Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Star | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031558.3 | NP_113746.1 | 6 - 7 | 859 | 109 | |
GenBank mRNA | AB001349.1 | - | 815 | 109 | ||
AB006007.1 | - | 6 - 7 | 860 | 109 | ||
AF044081.1 | - | 799 | 109 | |||
AY736357.1 | - | 6 - 7 | 740 | 109 | ||
BC088859.1 | - | 6 - 7 | 751 | 7311249 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |