Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ngb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033359.3 | NP_203523.2 | 2 - 3 | 613 | 88 | |
XM_008764864.1 | XP_008763086.1 | 2 - 3 | 1294 | 88 | ||
GenBank mRNA | AB056655.1 | - | 2 - 3 | 185 | 88 | |
AF333245.1 | - | 2 - 3 | 204 | 88 | ||
AF334379.1 | - | 2 - 3 | 204 | 88 | ||
AY066001.1 | - | 2 - 3 | 413 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Rattus norvegicus Probe spans exons |