Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Synm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134858.3 | NP_001128330.1 | 2 - 3 | 1158 | 96 | |
XM_006229337.2 | XP_006229399.1 | 2 - 3 | 1153 | 96 | ||
GenBank mRNA | AB091769.1 | - | 2 - 3 | 1066 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |