Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: URH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001180708.3 | NP_010688.4 | 1 | 551 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Saccharomyces cerevisiae Both primers and probe map within a single exon |