Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001180865.3 | NP_219497.4 | 1 - 2 | 49 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Saccharomyces cerevisiae Probe spans exons |