Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001182205.1 | NP_013420.2 | 2 - 3 | 164 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Saccharomyces cerevisiae Probe spans exons |