Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPS18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001183042.1 | NP_014195.1 | 1 | 854 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Saccharomyces cerevisiae Both primers and probe map within a single exon |