Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001183744.3 | NP_014969.3 | 1 | 1752 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Saccharomyces cerevisiae Both primers and probe map within a single exon |