Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001348754.1 | NP_001335683.1 | 4 - 5 | 902 | 86 | |
GenBank mRNA | AJ656918.1 | - | 1 - 2 | 98 | 86 |
Gene Symbol: LOC100622780 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021081948.1 | XP_020937607.1 | 939 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |