Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHTOP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243268.1 | NP_001230197.1 | 3 - 4 | 545 | 56 | |
GenBank mRNA | AK349088.1 | - | 3 - 4 | 502 | 56 | |
AK393545.1 | - | 3 - 4 | 525 | 56 | ||
EU814888.1 | - | 3 - 4 | 545 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |