Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214313.2 | NP_999478.1 | 4 - 5 | 263 | 128 | |
XM_021083263.1 | XP_020938922.1 | 460 | 128 | |||
GenBank mRNA | AF382821.1 | - | 4 - 5 | 263 | 128 | |
AK230709.1 | - | 4 - 5 | 324 | 128 | ||
AK347073.1 | - | 4 - 5 | 363 | 128 | ||
AK392783.1 | - | 4 - 5 | 383 | 128 | ||
AY609721.1 | - | 4 - 5 | 377 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Sus scrofa Inventoried Probe spans exons |