Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACYP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190198.1 | NP_001177127.1 | 2 - 3 | 413 | 74 | |
NM_001190199.1 | NP_001177128.1 | 2 - 3 | 173 | 74 | ||
XM_005656376.3 | XP_005656433.1 | 248 | 74 | |||
GenBank mRNA | AY609878.1 | - | 2 - 3 | 341 | 74 | |
CK452889.1 | - | 2 - 3 | 413 | 74 | ||
CX059140.1 | - | 1 - 2 | 115 | 74 | ||
EW062047.2 | - | 2 - 3 | 195 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Sus scrofa Probe spans exons |