Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NELFE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101825.1 | NP_001095295.1 | 8 - 9 | 967 | 90 | |
GenBank mRNA | AK233579.1 | - | 8 - 9 | 952 | 90 | |
AK236334.1 | - | 8 - 9 | 1091 | 90 | ||
AK389553.1 | - | 8 - 9 | 977 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |