Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFEMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244261.1 | NP_001231190.1 | 10 - 11 | 1460 | 110 | |
XM_013996146.2 | XP_013851600.1 | 1336 | 110 | |||
GenBank mRNA | HQ630067.1 | - | 10 - 11 | 1480 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Sus scrofa Probe spans exons |