Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021089236.1 | XP_020944895.1 | 1792 | 96 | ||
GenBank mRNA | AB042526.1 | - | 1 | 46 | 96 | |
AF327369.1 | - | 1 | 442 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |