Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEBPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003127015.4 | XP_003127063.1 | 1144 | 61 | ||
GenBank mRNA | AF103944.1 | - | 1 | 152 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Inventoried Both primers and probe map within a single exon |