Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAPPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001926753.7 | XP_001926788.5 | 2551 | 71 | ||
GenBank mRNA | AF421142.1 | - | 2 - 3 | 360 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |