Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005672249.3 | XP_005672306.1 | 2461 | 87 | ||
GenBank mRNA | AY079213.1 | - | 1 | 446 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |